RIE156322824 Rieke Metals, Inc.

3'-chloro-2,2-dimethylbutyrophenone

  • MDL number MFCD03841132
  • Cas Number : 898765-40-7
  • Molecular Weight : 210.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6121-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-20
Availability details for
Quantity
  • Availability
  • Add to Cart