RIE156322825 Rieke Metals, Inc.

4'-chloro-2,2-dimethylbutyrophenone

  • MDL number MFCD03841133
  • Cas Number : 898765-43-0
  • Molecular Weight : 210.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6121-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-21
Availability details for
Quantity
  • Availability
  • Add to Cart