RIE156322828 Rieke Metals, Inc.

2',2,2,3'-Tetramethylbutyrophenone

  • MDL number MFCD03841136
  • Cas Number : 898765-52-1
  • Molecular Weight : 204.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6121-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-26
Availability details for
Quantity
  • Availability
  • Add to Cart