RIE156322830 Rieke Metals, Inc.

2',2,2,5'-Tetramethylbutyrophenone

  • MDL number MFCD03841138
  • Cas Number : 898765-58-7
  • Molecular Weight : 204.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6121-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-28
Availability details for
Quantity
  • Availability
  • Add to Cart