RIE156322832 Rieke Metals, Inc.

2,2,3',4'-tetramethylbutyrophenone

  • MDL number MFCD03841140
  • Cas Number : 898765-64-5
  • Molecular Weight : 204.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3,4-dimethylphenyl)-2,2-dimethyl-1-butanone
Inchi Code 1S/C14H20O/c1-6-14(4,5)13(15)12-8-7-10(2)11(3)9-12/h7-9H,6H2,1-5H3
Inchi Key NYTPFGDALUCTBM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6121-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-30
Availability details for
Quantity
  • Availability
  • Add to Cart