RIE156322834 Rieke Metals, Inc.

4'-bromo-2,2-dimethyl-3'-fluorobutyrophenone

  • MDL number MFCD07699550
  • Cas Number : 898765-68-9
  • Molecular Weight : 273.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6121-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-32
Availability details for
Quantity
  • Availability
  • Add to Cart