RIE156322836 Rieke Metals, Inc.

3'-chloro-2,2-dimethyl-4'-fluorobutyrophenone

  • MDL number MFCD07699552
  • Cas Number : 898765-72-5
  • Molecular Weight : 228.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6121-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-34
Availability details for
Quantity
  • Availability
  • Add to Cart