RIE156322837 Rieke Metals, Inc.

2'-Chloro-2,2-dimethylbutyrophenone

  • MDL number MFCD07699553
  • Cas Number : 898765-74-7
  • Molecular Weight : 210.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-chlorophenyl)-2,2-dimethyl-1-butanone
Inchi Code 1S/C12H15ClO/c1-4-12(2,3)11(14)9-7-5-6-8-10(9)13/h5-8H,4H2,1-3H3
Inchi Key UVVPBMYQGFOWTL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6121-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-36
Availability details for
Quantity
  • Availability
  • Add to Cart