RIE156322838 Rieke Metals, Inc.

2,2-dimethyl-2'-fluorobutyrophenone

  • MDL number MFCD03841146
  • Cas Number : 898765-76-9
  • Molecular Weight : 194.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6121-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-37
Availability details for
Quantity
  • Availability
  • Add to Cart