RIE156322844 Rieke Metals, Inc.

3'-Chloro-2,2-dimethyl-5'-fluorobutyrophenone

  • MDL number MFCD03841152
  • Cas Number : 898765-88-3
  • Molecular Weight : 228.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6121-51

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-51
Availability details for
Quantity
  • Availability
  • Add to Cart