RIE156322849 Rieke Metals, Inc.

3',4'-dichloro-2,2-dimethylbutyrophenone

  • MDL number MFCD03841157
  • Cas Number : 898765-98-5
  • Molecular Weight : 245.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6121-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-56
Availability details for
Quantity
  • Availability
  • Add to Cart