RIE156322851 Rieke Metals, Inc.

2',4'-difluoro-2,2-dimethylbutyrophenone

  • MDL number MFCD03841159
  • Cas Number : 898766-02-4
  • Molecular Weight : 212.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6121-58

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-58
Availability details for
Quantity
  • Availability
  • Add to Cart