RIE156322852 Rieke Metals, Inc.

3',4'-Difluoro-2,2-dimethylbutyrophenone

  • MDL number MFCD03841160
  • Cas Number : 898766-04-6
  • Molecular Weight : 212.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3,4-difluorophenyl)-2,2-dimethyl-1-butanone
Inchi Code 1S/C12H14F2O/c1-4-12(2,3)11(15)8-5-6-9(13)10(14)7-8/h5-7H,4H2,1-3H3
Inchi Key RWMWTQKVMXXHAI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6121-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-59
Availability details for
Quantity
  • Availability
  • Add to Cart