RIE156322853 Rieke Metals, Inc.

3',5'-difluoro-2,2-dimethylbutyrophenone

  • MDL number MFCD03841161
  • Cas Number : 898766-06-8
  • Molecular Weight : 212.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6121-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6121-60
Availability details for
Quantity
  • Availability
  • Add to Cart