RIE156322858 Rieke Metals, Inc.

4'-Cyano-2,2-dimethylpropiophenone

  • MDL number MFCD03841169
  • Cas Number : 150009-08-8
  • Molecular Weight : 187.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2,2-dimethylpropanoyl)benzonitrile
Inchi Code 1S/C12H13NO/c1-12(2,3)11(14)10-6-4-9(8-13)5-7-10/h4-7H,1-3H3
Inchi Key FCXVFMKJNOPZLF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6122-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6122-10
Availability details for
Quantity
  • Availability
  • Add to Cart