RIE156322860 Rieke Metals, Inc.

3'-Carboethoxy-2,2-dimethylpropiophenone

  • MDL number MFCD03841171
  • Cas Number : 898766-18-2
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6122-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6122-12
Availability details for
Quantity
  • Availability
  • Add to Cart