RIE156322864 Rieke Metals, Inc.

3'-Chloro-2,2-dimethylpropiophenone

  • MDL number MFCD03841176
  • Cas Number : 53226-55-4
  • Molecular Weight : 196.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6122-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6122-20
Availability details for
Quantity
  • Availability
  • Add to Cart