RIE156322868 Rieke Metals, Inc.

2,2,3',5'-tetramethylpropiophenone

  • MDL number MFCD03841184
  • Cas Number : 898766-24-0
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6122-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6122-31
Availability details for
Quantity
  • Availability
  • Add to Cart