RIE156322870 Rieke Metals, Inc.

4'-chloro-2,2-dimethyl-3'-fluoropropiophenone

  • MDL number MFCD07699555
  • Cas Number : 898766-30-8
  • Molecular Weight : 214.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6122-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6122-33
Availability details for
Quantity
  • Availability
  • Add to Cart