RIE156322874 Rieke Metals, Inc.

2,2-dimethyl-2'-trifluoromethylpropiophenone

  • MDL number MFCD03841190
  • Cas Number : 898766-36-4
  • Molecular Weight : 230.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6122-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6122-43
Availability details for
Quantity
  • Availability
  • Add to Cart