RIE156322876 Rieke Metals, Inc.

2,2-dimethyl-4'-trifluoromethylpropiophenone

  • MDL number MFCD03841192
  • Cas Number : 586346-65-8
  • Molecular Weight : 230.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6122-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6122-45
Availability details for
Quantity
  • Availability
  • Add to Cart