RIE156322879 Rieke Metals, Inc.

3'-chloro-2,2-dimethyl-5'-fluoropropiophenone

  • MDL number MFCD03841195
  • Cas Number : 898766-48-8
  • Molecular Weight : 214.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6122-51

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6122-51
Availability details for
Quantity
  • Availability
  • Add to Cart