RIE156322881 Rieke Metals, Inc.

2',3'-dichloro-2,2-dimethylpropiophenone

  • MDL number MFCD03841197
  • Cas Number : 898766-54-6
  • Molecular Weight : 231.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6122-53

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6122-53
Availability details for
Quantity
  • Availability
  • Add to Cart