RIE156322884 Rieke Metals, Inc.

3',5'-Dichloro-2,2-dimethylpropiophenone

  • MDL number MFCD03841201
  • Cas Number : 898766-63-7
  • Molecular Weight : 231.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3,5-dichlorophenyl)-2,2-dimethyl-1-propanone
Inchi Code 1S/C11H12Cl2O/c1-11(2,3)10(14)7-4-8(12)6-9(13)5-7/h4-6H,1-3H3
Inchi Key VFRLXTNREOXDJW-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6122-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6122-57
Availability details for
Quantity
  • Availability
  • Add to Cart