RIE156322888 Rieke Metals, Inc.

2,2-dimethyl-3',4',5'-trifluoropropiophenone

  • MDL number MFCD03841205
  • Cas Number : 898789-80-5
  • Molecular Weight : 216.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6122-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6122-61
Availability details for
Quantity
  • Availability
  • Add to Cart