RIE156322890 Rieke Metals, Inc.

Cyclopropyl 3-methylphenyl ketone

  • MDL number MFCD03841207
  • Cas Number : 150668-37-4
  • Molecular Weight : 160.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-01-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-01-03
Availability details for
Quantity
  • Availability
  • Add to Cart