RIE156322896 Rieke Metals, Inc.

4-Carboethoxyphenyl cyclopropyl ketone

  • MDL number MFCD03841215
  • Cas Number : 863769-67-9
  • Molecular Weight : 218.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-(cyclopropylcarbonyl)benzoate
Inchi Code 1S/C13H14O3/c1-2-16-13(15)11-7-5-10(6-8-11)12(14)9-3-4-9/h5-9H,2-4H2,1H3
Inchi Key XSMZWMTZXKUJKE-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-01-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-01-13
Availability details for
Quantity
  • Availability
  • Add to Cart