RIE156322901 Rieke Metals, Inc.

Cyclopropyl 3-fluorophenyl ketone

  • MDL number MFCD03841220
  • Cas Number : 77972-82-8
  • Molecular Weight : 164.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-01-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-01-22
Availability details for
Quantity
  • Availability
  • Add to Cart