RIE156322902 Rieke Metals, Inc.

cyclopropyl 2,3-dimethylphenyl ketone

  • MDL number MFCD03841221
  • Cas Number : 898790-00-6
  • Molecular Weight : 174.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-01-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-01-26
Availability details for
Quantity
  • Availability
  • Add to Cart