RIE156322905 Rieke Metals, Inc.

4-Bromo-3-fluorophenyl cyclopropyl ketone

  • MDL number MFCD03841224
  • Cas Number : 898790-03-9
  • Molecular Weight : 243.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromo-3-fluorophenyl)(cyclopropyl)methanone
Inchi Code 1S/C10H8BrFO/c11-8-4-3-7(5-9(8)12)10(13)6-1-2-6/h3-6H,1-2H2
Inchi Key FPVIKFLEKLICRG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-01-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-01-32
Availability details for
Quantity
  • Availability
  • Add to Cart