RIE156322906 Rieke Metals, Inc.

4-Chloro-3-fluorophenyl cyclopropyl ketone

  • MDL number MFCD03841225
  • Cas Number : 898790-06-2
  • Molecular Weight : 198.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-01-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-01-33
Availability details for
Quantity
  • Availability
  • Add to Cart