RIE156322907 Rieke Metals, Inc.

3-Chloro-4-fluorophenyl cyclopropyl ketone

  • MDL number MFCD03841226
  • Cas Number : 898790-09-5
  • Molecular Weight : 198.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chloro-4-fluorophenyl)(cyclopropyl)methanone
Inchi Code 1S/C10H8ClFO/c11-8-5-7(3-4-9(8)12)10(13)6-1-2-6/h3-6H,1-2H2
Inchi Key UWLYSHCUFNFEPS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-01-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-01-34
Availability details for
Quantity
  • Availability
  • Add to Cart