RIE156322909 Rieke Metals, Inc.

2-Fluorophenyl cyclopropyl ketone

  • MDL number MFCD03841228
  • Cas Number : 141030-72-0
  • Molecular Weight : 164.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-01-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-01-37
Availability details for
Quantity
  • Availability
  • Add to Cart