RIE156322910 Rieke Metals, Inc.

Cyclopropyl 2-trifluoromethylphenyl ketone

  • MDL number MFCD03841229
  • Cas Number : 898790-12-0
  • Molecular Weight : 214.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclopropyl[2-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C11H9F3O/c12-11(13,14)9-4-2-1-3-8(9)10(15)7-5-6-7/h1-4,7H,5-6H2
Inchi Key GLQVXMURVDLEDP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-01-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-01-43
Availability details for
Quantity
  • Availability
  • Add to Cart