RIE156322912 Rieke Metals, Inc.

4-Bromo-2-fluorophenyl cyclopropyl ketone

  • MDL number MFCD03841232
  • Cas Number : 898790-15-3
  • Molecular Weight : 243.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-01-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-01-49
Availability details for
Quantity
  • Availability
  • Add to Cart