RIE156322913 Rieke Metals, Inc.

2-Chloro-4-fluorophenyl cyclopropyl ketone

  • MDL number MFCD03841233
  • Cas Number : 898790-18-6
  • Molecular Weight : 198.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-01-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-01-50
Availability details for
Quantity
  • Availability
  • Add to Cart