RIE156322919 Rieke Metals, Inc.

Cyclopropyl 3,5-dichlorophenyl ketone

  • MDL number MFCD03841240
  • Cas Number : 898790-30-2
  • Molecular Weight : 215.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-01-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-01-57
Availability details for
Quantity
  • Availability
  • Add to Cart