RIE156322924 Rieke Metals, Inc.

cyclobutyl 3-methylphenyl ketone

  • MDL number MFCD03841246
  • Cas Number : 898790-40-4
  • Molecular Weight : 174.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-02-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-03
Availability details for
Quantity
  • Availability
  • Add to Cart