RIE156322925 Rieke Metals, Inc.

Cyclobutyl 2-methoxyphenyl ketone

  • MDL number MFCD03841248
  • Cas Number : 898790-42-6
  • Molecular Weight : 190.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclobutyl(2-methoxyphenyl)methanone
Inchi Code 1S/C12H14O2/c1-14-11-8-3-2-7-10(11)12(13)9-5-4-6-9/h2-3,7-9H,4-6H2,1H3
Inchi Key VRZXSSDRRKBMRP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-02-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-05
Availability details for
Quantity
  • Availability
  • Add to Cart