RIE156322931 Rieke Metals, Inc.

2-Carboethoxyphenyl cyclobutyl ketone

  • MDL number MFCD03841254
  • Cas Number : 898790-52-8
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-02-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-11
Availability details for
Quantity
  • Availability
  • Add to Cart