RIE156322932 Rieke Metals, Inc.

3-carboethoxyphenyl cyclobutyl ketone

  • MDL number MFCD03841255
  • Cas Number : 898790-54-0
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-02-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-12
Availability details for
Quantity
  • Availability
  • Add to Cart