RIE156322934 Rieke Metals, Inc.

cyclobutyl 2-thiomethylphenyl ketone

  • MDL number MFCD03841257
  • Cas Number : 898790-56-2
  • Molecular Weight : 206.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-02-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-14
Availability details for
Quantity
  • Availability
  • Add to Cart