RIE156322935 Rieke Metals, Inc.

Cyclobutyl 4-thiomethylphenyl ketone

  • MDL number MFCD03841258
  • Cas Number : 716341-27-4
  • Molecular Weight : 206.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-02-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-15
Availability details for
Quantity
  • Availability
  • Add to Cart