RIE156322936 Rieke Metals, Inc.

3-bromophenyl cyclobutyl ketone

  • MDL number MFCD03841259
  • Cas Number : 898790-58-4
  • Molecular Weight : 239.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-02-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-18
Availability details for
Quantity
  • Availability
  • Add to Cart