RIE156322938 Rieke Metals, Inc.

3-Chlorophenyl cyclobutyl ketone

  • MDL number MFCD03841261
  • Cas Number : 898790-62-0
  • Molecular Weight : 194.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-02-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-20
Availability details for
Quantity
  • Availability
  • Add to Cart