RIE156322941 Rieke Metals, Inc.

Cyclobutyl 2,3-dimethylphenyl ketone

  • MDL number MFCD03841264
  • Cas Number : 898790-66-4
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-02-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-26
Availability details for
Quantity
  • Availability
  • Add to Cart