RIE156322942 Rieke Metals, Inc.

cyclobutyl 2,4-dimethylphenyl ketone

  • MDL number MFCD03841265
  • Cas Number : 898790-68-6
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclobutyl(2,4-dimethylphenyl)methanone
Inchi Code 1S/C13H16O/c1-9-6-7-12(10(2)8-9)13(14)11-4-3-5-11/h6-8,11H,3-5H2,1-2H3
Inchi Key JZCNVUQXOTVPQU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6123-02-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-27
Availability details for
Quantity
  • Availability
  • Add to Cart