RIE156322945 Rieke Metals, Inc.

cyclobutyl 3,4-dimethylphenyl ketone

  • MDL number MFCD03841268
  • Cas Number : 898790-76-6
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-02-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-30
Availability details for
Quantity
  • Availability
  • Add to Cart