RIE156322947 Rieke Metals, Inc.

4-Bromo-3-fluorophenyl cyclobutyl ketone

  • MDL number MFCD03841270
  • Cas Number : 898790-82-4
  • Molecular Weight : 257.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-02-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-32
Availability details for
Quantity
  • Availability
  • Add to Cart