RIE156322948 Rieke Metals, Inc.

4-chloro-3-fluorophenyl cyclobutyl ketone

  • MDL number MFCD03841271
  • Cas Number : 898790-85-7
  • Molecular Weight : 212.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6123-02-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6123-02-33
Availability details for
Quantity
  • Availability
  • Add to Cart